Diethyl maleate, 97%
Diethyl maleate, 97%
Diethyl maleate, 97%
Thermo Scientific Chemicals

Diethyl maleate, 97%

CAS: 141-05-9 | C8H12O4 | 172.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1000 g
Catalog number L03876.06
also known as L03876-06
Price (USD)
23.65
Online exclusive
26.20
Save 2.55
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
23.65
Online exclusive
26.20
Save 2.55
Each
Chemical Identifiers
CAS141-05-9
IUPAC Name1,4-diethyl (2Z)-but-2-enedioate
Molecular FormulaC8H12O4
InChI KeyIEPRKVQEAMIZSS-WAYWQWQTSA-N
SMILESCCOC(=O)\C=C/C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid or viscous liquid
Identification (FTIR)Conforms
Refractive Index1.4380-1.4430 @ 20?C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only