Dimethoxydiphenylsilane, 97%
Dimethoxydiphenylsilane, 97%
Dimethoxydiphenylsilane, 97%
Thermo Scientific Chemicals

Dimethoxydiphenylsilane, 97%

CAS: 6843-66-9 | C14H16O2Si | 244.365 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number L04031.30
also known as L04031-30
Price (USD)
144.65
Special offer
Online exclusive
Ends: 30-Jun-2026
170.00
Save 25.35 (15%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
144.65
Special offer
Online exclusive
Ends: 30-Jun-2026
170.00
Save 25.35 (15%)
Each
Chemical Identifiers
CAS6843-66-9
IUPAC Namedimethoxydiphenylsilane
Molecular FormulaC14H16O2Si
InChI KeyAHUXYBVKTIBBJW-UHFFFAOYSA-N
SMILESCO[Si](OC)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥96.0% (non-U.S. sourced material)
CommentMaterial Sourced in the U.S. and in other countries
Refractive Index1.5395-1.5445 @ 20°C (non-U.S. sourced material)
Assay from Supplier's CofA≥96.0% (U.S. sourced material)
View more
Reagent for preparation of diphenylsilylene derivatives of diols by exchange.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only