Vinyl(chloromethyl)dimethylsilane, 97%
Vinyl(chloromethyl)dimethylsilane, 97%
Vinyl(chloromethyl)dimethylsilane, 97%
Thermo Scientific Chemicals

Vinyl(chloromethyl)dimethylsilane, 97%

CAS: 16709-86-7 | C5H11ClSi | 134.678 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L04056.14
also known as L04056-14
Price (USD)
237.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
237.00
Each
Chemical Identifiers
CAS16709-86-7
IUPAC Name(chloromethyl)(ethenyl)dimethylsilane
Molecular FormulaC5H11ClSi
InChI KeySZZZMXFBEKWPBU-UHFFFAOYSA-N
SMILESC[Si](C)(CCl)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0% (non-U.S. Specification)
CommentPurchased in the U.S. and in other countries
FormLiquid
Appearance (Color)Clear colorless to yellow
Assay from Supplier's CofA≥96.0% (GC) (U.S. Specification)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only