1,2-Bis(phenylsulfonyl)ethane, 98+%
1,2-Bis(phenylsulfonyl)ethane, 98+%
1,2-Bis(phenylsulfonyl)ethane, 98+%
Thermo Scientific Chemicals

1,2-Bis(phenylsulfonyl)ethane, 98+%

CAS: 599-94-0 | C14H14O4S2 | 310.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L04083.06
also known as L04083-06
Price (USD)
70.65
Online Exclusive
78.40
Save 7.75 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
70.65
Online Exclusive
78.40
Save 7.75 (10%)
Each
Chemical Identifiers
CAS599-94-0
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥98.0%
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only