Methyldiphenylsilane, 97%
Methyldiphenylsilane, 97%
Methyldiphenylsilane, 97%
Thermo Scientific Chemicals

Methyldiphenylsilane, 97%

CAS: 776-76-1 | C13H13Si | 197.332 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L04211.06
also known as L04211-06
Price (USD)
42.80
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
42.80
Each
Chemical Identifiers
CAS776-76-1
IUPAC Namemethyldiphenylsilyl
Molecular FormulaC13H13Si
InChI KeyOKHRRIGNGQFVEE-UHFFFAOYSA-N
SMILESC[Si](C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
FormLiquid
Assay from Suppliers CofA≥96%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only