3-Piperidinemethanol, 96%
3-Piperidinemethanol, 96%
3-Piperidinemethanol, 96%
Thermo Scientific Chemicals

3-Piperidinemethanol, 96%

CAS: 4606-65-9 | C6H13NO | 115.176 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L04264.14
also known as L04264-14
Price (USD)
120.65
Special offer
Online exclusive
Ends: 30-Jun-2026
142.00
Save 21.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
120.65
Special offer
Online exclusive
Ends: 30-Jun-2026
142.00
Save 21.35 (15%)
Each
Chemical Identifiers
CAS4606-65-9
IUPAC Name(3R)-3-(hydroxymethyl)piperidin-1-ium
Molecular FormulaC6H14NO
InChI KeyVUNPWIPIOOMCPT-ZCFIWIBFSA-O
SMILESOC[C@@H]1CCC[NH2+]C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)56.0-66.0?C
Appearance (Color)Colorless to white to cream to yellow to orange
FormCrystals or powder or crystalline powder or fused solid or clear liquid or viscous liquid as melt
Assay (GC)≥95.0%
Water Content (Karl Fischer Titration)≤2%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only