2-Amino-4'-bromoacetophenone hydrochloride, 98%
2-Amino-4'-bromoacetophenone hydrochloride, 98%
2-Amino-4'-bromoacetophenone hydrochloride, 98%
Thermo Scientific Chemicals

2-Amino-4'-bromoacetophenone hydrochloride, 98%

CAS: 5467-72-1 | C8H9BrClNO | 250.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number L04292.09
also known as L04292-09
Price (USD)
164.65
Special offer
Online exclusive
Ends: 30-Jun-2026
193.00
Save 28.35 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
164.65
Special offer
Online exclusive
Ends: 30-Jun-2026
193.00
Save 28.35 (15%)
Each
Chemical Identifiers
CAS5467-72-1
IUPAC Name2-(4-bromophenyl)-2-oxoethan-1-aminium chloride
Molecular FormulaC8H9BrClNO
InChI KeyROAVTVXTYFSQEA-UHFFFAOYSA-N
SMILES[Cl-].[NH3+]CC(=O)C1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥97.5%
Appearance (Color)White to cream to pink to orange to pale brown
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only