(1-Tetradecyl)triphenylphosphonium bromide, 97%
(1-Tetradecyl)triphenylphosphonium bromide, 97%
(1-Tetradecyl)triphenylphosphonium bromide, 97%
Thermo Scientific Chemicals

(1-Tetradecyl)triphenylphosphonium bromide, 97%

CAS: 25791-20-2 | C32H44BrP | 539.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L04311.22
also known as L04311-22
Price (USD)
220.65
Special offer
Online exclusive
Ends: 30-Jun-2026
259.00
Save 38.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
220.65
Special offer
Online exclusive
Ends: 30-Jun-2026
259.00
Save 38.35 (15%)
Each
Chemical Identifiers
CAS25791-20-2
IUPAC Nametriphenyl(tetradecyl)phosphanium bromide
Molecular FormulaC32H44BrP
InChI KeyFUMBGFNGBMYHGH-UHFFFAOYSA-M
SMILES[Br-].CCCCCCCCCCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or blue
FormCrystals or powder or crystalline powder or lumps or flakes
Assay (Titration ex Bromide)≥96.0 to ≤104.0% (non-U.S. sourced material)
Assay from Supplier's CofA≥96.0% (U.S. sourced material)
CommentMaterial sourced in the U.S. and in other countries
View more
(1-Tetradecyl)triphenylphosphonium bromide is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Tetradecyl)triphenylphosphonium bromide is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Slightly soluble in Water.

Notes
Hygroscopic. Store away from oxidizing agents. Incompatible with moisture and oxidizing agents.
RUO – Research Use Only