6-Ethoxy-2-mercaptobenzothiazole, 99%
6-Ethoxy-2-mercaptobenzothiazole, 99%
6-Ethoxy-2-mercaptobenzothiazole, 99%
6-Ethoxy-2-mercaptobenzothiazole, 99%
Thermo Scientific Chemicals

6-Ethoxy-2-mercaptobenzothiazole, 99%

CAS: 120-53-6 | C9H9NOS2 | 211.297 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L04447.14
also known as L04447-14
Price (USD)
299.65
Online Exclusive
333.00
Save 33.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
299.65
Online Exclusive
333.00
Save 33.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS120-53-6
IUPAC Name6-ethoxy-2,3-dihydro-1,3-benzothiazole-2-thione
Molecular FormulaC9H9NOS2
InChI KeyHOASVNMVYBSLSU-UHFFFAOYSA-N
SMILESCCOC1=CC=C2NC(=S)SC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to pale brown
FormCrystals or powder or crystalline powder
Assay (HPLC)≥98.5%
Melting Point (clear melt)196.0-202.0?C
6-Ethoxy-2-mercaptobenzothiazole is used as a reagent in the preparation of nitrothiazolythio benzothiazoles as carbon-junction N-terminal kinase inhibitors. It is used in the synthesis of active pharmaceutical ingredients. Further, it serves as a matrix-assisted laser desorption mass spectrometry for bio molecules mass determination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Ethoxy-2-mercaptobenzothiazole is used as a reagent in the preparation of nitrothiazolythio benzothiazoles as carbon-junction N-terminal kinase inhibitors. It is used in the synthesis of active pharmaceutical ingredients. Further, it serves as a matrix-assisted laser desorption mass spectrometry for bio molecules mass determination.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong bases and halogens.
RUO – Research Use Only

General References:

  1. Joshi, A.; Nagaiah, T. C. Nitrogen-doped carbon nanotubes for sensitive and selective determination of heavy metals. RSC Adv. 2015, 5 (127), 105119-105127.
  2. Dong, J.; Fang, Q.; He, H.; Zhang, Y.; Xu, J.; Sun, Y. Electrochemical sensor based on EDTA intercalated into layered double hydroxides of magnesium and aluminum for ultra trace level detection of lead(II). Microchim. Acta 2015, 182 (3), 653-659.