2-Picoline N-oxide, 98%
2-Picoline N-oxide, 98%
2-Picoline N-oxide, 98%
Thermo Scientific Chemicals

2-Picoline N-oxide, 98%

CAS: 931-19-1 | C6H7NO | 109.128 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number L04488.14
also known as L04488-14
Price (USD)
27.65
Special offer
Online exclusive
Ends: 30-Jun-2026
32.70
Save 5.05 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
27.65
Special offer
Online exclusive
Ends: 30-Jun-2026
32.70
Save 5.05 (15%)
Each
Chemical Identifiers
CAS931-19-1
IUPAC Name2-methylpyridin-1-ium-1-olate
Molecular FormulaC6H7NO
InChI KeyCFZKDDTWZYUZKS-UHFFFAOYSA-N
SMILESCC1=CC=CC=[N+]1[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFused solid or liquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Appearance (Color)White to cream or pale yellow or pale brown to very dark brown
2-Picoline N-oxide is used in proteomics research. Also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Picoline N-oxide is used in proteomics research. Also used as pharmaceutical intermediates.

Solubility
Soluble in water.

Notes
Hygroscopic. Store under dry inert gas. Incompatible with acids, Acid chlorides, Acid anhydrides, Chloroformates.
RUO – Research Use Only

General References:

  1. Shigeru. Oae; Teijiro. Kitao; Yoshinori. Kitaoka. The Mechanism of the Reaction of 2-Picoline N-Oxide with Acetic Anhydride.J. Am. Chem. Soc.1962, 84 (17), 3359-3362.
  2. R. S. Sagerr; W. H. Watson. 2-Picoline N-oxide complexes of copper(II)chloride. The crystal structure of Cu3 Cl6(C6H7NO)2.2H2O.Inorg. Chem.1968, 7 (10), 2035-2040.