4-Chlorotoluene, 98%
4-Chlorotoluene, 98%
4-Chlorotoluene, 98%
Thermo Scientific Chemicals

4-Chlorotoluene, 98%

CAS: 106-43-4 | C7H7Cl | 126.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1000 g
Catalog number L04672.0B
also known as L04672-0B
Price (USD)
136.65
Special offer
Online exclusive
Ends: 30-Jun-2026
160.00
Save 23.35 (15%)
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
136.65
Special offer
Online exclusive
Ends: 30-Jun-2026
160.00
Save 23.35 (15%)
Each
Chemical Identifiers
CAS106-43-4
IUPAC Name1-chloro-4-methylbenzene
Molecular FormulaC7H7Cl
InChI KeyNPDACUSDTOMAMK-UHFFFAOYSA-N
SMILESCC1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive Index1.5190-1.5230 @ 20?C
FormLiquid
Assay (GC)≥97.5%
4-Chlorotoluene is used for the preparation model gas of 4-chlorotoulene by passing synthesized air into liquid 4-chlorotoulene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorotoluene is used for the preparation model gas of 4-chlorotoulene by passing synthesized air into liquid 4-chlorotoulene.

Solubility
Soluble in water (0.04 g/L at 20oc).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Zhang C.; Trudell ML. Palladium-bisimidazol-2-ylidene complexes as catalysts for general and efficient Suzuki cross-coupling reactions of aryl chlorides with arylboronic acids. Tetrahedron Lett. 2000, 41 (5), 595-598.
  2. Maria M. Palaiologou. Densities, Viscosities, and Refractive Indices of Some Alkyl Esters with 4-Chlorotoluene Systems at (293.15, 298.15, and 303.15) K. J. Chem. Eng. Data. 1996, 41 (5), 1036-1039.