(+/-)-Mandelamide, 97%
(+/-)-Mandelamide, 97%
(+/-)-Mandelamide, 97%
Thermo Scientific Chemicals

(+/-)-Mandelamide, 97%

CAS: 4410-31-5 | C8H9NO2 | 151.165 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L04679.06
also known as L04679-06
Price (USD)
122.65
Special offer
Online exclusive
Ends: 30-Jun-2026
144.00
Save 21.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
122.65
Special offer
Online exclusive
Ends: 30-Jun-2026
144.00
Save 21.35 (15%)
Each
Chemical Identifiers
CAS4410-31-5
IUPAC Name2-hydroxy-2-phenylacetamide
Molecular FormulaC8H9NO2
InChI KeyMAGPZHKLEZXLNU-UHFFFAOYNA-N
SMILESNC(=O)C(O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Appearance (Color)White to pale cream
Melting Point (clear melt)130.5-136.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only