Phenyltriethoxysilane, 98%
Phenyltriethoxysilane, 98%
Phenyltriethoxysilane, 98%
Thermo Scientific Chemicals

Phenyltriethoxysilane, 98%

CAS: 780-69-8 | C12H20O3Si | 240.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number L04684.0B
also known as L04684-0B
Price (USD)
126.65
Special offer
Online exclusive
Ends: 30-Jun-2026
149.00
Save 22.35 (15%)
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
126.65
Special offer
Online exclusive
Ends: 30-Jun-2026
149.00
Save 22.35 (15%)
Each
Chemical Identifiers
CAS780-69-8
IUPAC Nametriethoxy(phenyl)silane
Molecular FormulaC12H20O3Si
InChI KeyJCVQKRGIASEUKR-UHFFFAOYSA-N
SMILESCCO[Si](OCC)(OCC)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)Clear colorless
Refractive Index1.4590-1.4630 @ 20?C
FormLiquid or viscous liquid
Identification (FTIR)Conforms
Phenyltriethoxysilane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyltriethoxysilane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture. Store under dry inert gas.
RUO – Research Use Only

General References:

  1. M. M. Sprung.; F. O. Guenther. The hydrolysis of n-amyltriethoxysilane and phenyltriethoxysilane. Journal of Polymer Science. 1958, 28 (116), 17-34.
  2. Zhijian Wu.; Hong Xiang.; Taehoon Kim.; Myung-Suk Chun.; Kangtaek Lee. Surface properties of submicrometer silica spheres modified with aminopropyltriethoxysilane and phenyltriethoxysilane. Journal of Colloid and Interface Science. 2006, 304 (1), 119-124.