Trimethylamine hydrochloride, 98%
Trimethylamine hydrochloride, 98%
Trimethylamine hydrochloride, 98%
Thermo Scientific Chemicals

Trimethylamine hydrochloride, 98%

CAS: 593-81-7 | C3H10ClN | 95.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number L04718.30
also known as L04718-30
Price (USD)
60.65
Special offer
Online exclusive
Ends: 30-Jun-2026
70.90
Save 10.25 (14%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
60.65
Special offer
Online exclusive
Ends: 30-Jun-2026
70.90
Save 10.25 (14%)
Each
Chemical Identifiers
CAS593-81-7
IUPAC Namehydrogen trimethylamine chloride
Molecular FormulaC3H10ClN
InChI KeySZYJELPVAFJOGJ-UHFFFAOYSA-N
SMILES[H+].[Cl-].CN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
FormMoist crystals or crystalline powder or powder
Assay (Titration ex Chloride)≥97.5 to ≤102.5% (Dry wt. basis)
Appearance (Color)White
Trimethylamine hydrochloride is used in the manufacture of quaternary ammonium compounds, as insect attractant. It is used as a raw material for the synthesis of choline chloride, quaternary ammonium compounds, ion exchange resins. It acts as cationic starch reagents, phase transfer catalyst, oil field chemicals, warning agent for natural gas and flotation agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylamine hydrochloride is used in the manufacture of quaternary ammonium compounds, as insect attractant. It is used as a raw material for the synthesis of choline chloride, quaternary ammonium compounds, ion exchange resins. It acts as cationic starch reagents, phase transfer catalyst, oil field chemicals, warning agent for natural gas and flotation agent.

Solubility
Soluble in water (737 mg/ml at 20°C), chloroform (moderately), and alcohol. Insoluble in ether.

Notes
Hygroscopic. Store under dry inert gas and away from moisture. Incompatible with oxidizing agents and water.
RUO – Research Use Only

General References:

  1. Smaranda Craciun; Emily P Balskus. Microbial conversion of choline to trimethylamine requires a glycyl radical enzyme.PNAS.2007, 109 (52), 21307-21312.
  2. Masao Miyake; Hiroshi Motonami; Suguru Shiomi; Tetsuji Hirato. Electrodeposition of purified aluminum coatings from dimethylsulfone-AlCl3 electrolytes with trimethylamine hydrochloride..Surface and Coatings Technology.2012, 206 4225-4229.