(Ethylthio)acetic acid, 97%
(Ethylthio)acetic acid, 97%
(Ethylthio)acetic acid, 97%
Thermo Scientific Chemicals

(Ethylthio)acetic acid, 97%

CAS: 627-04-3 | C4H8O2S | 120.166 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L04743.06
also known as L04743-06
Price (USD)
40.65
Online Exclusive
44.50
Save 3.85 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
40.65
Online Exclusive
44.50
Save 3.85 (9%)
Each
Chemical Identifiers
CAS627-04-3
IUPAC Name2-(ethylsulfanyl)acetic acid
Molecular FormulaC4H8O2S
InChI KeyVJIKFWJCVWFZIN-UHFFFAOYSA-N
SMILESCCSCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless to pale yellow
Assay (Aqueous acid-base Titration)>96.0 to <104.0%
Assay (Silylated GC)>96.0%
Refractive Index1.4830-1.4890 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only