Allyl phenyl sulfide, 97%, Thermo Scientific Chemicals
Allyl phenyl sulfide, 97%, Thermo Scientific Chemicals
Allyl phenyl sulfide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Allyl phenyl sulfide, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
L04771.09
also known as L04771-09
10 g
Catalog number L04771.09
also known as L04771-09
Price (USD)
68.65
Online exclusive
76.50
Save 7.85
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
68.65
Online exclusive
76.50
Save 7.85
Each
Chemical Identifiers
CAS5296-64-0
IUPAC Name(prop-2-en-1-ylsulfanyl)benzene
Molecular FormulaC9H10S
InChI KeyQGNRLAFFKKBSIM-UHFFFAOYSA-N
SMILESC=CCSC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5715-1.5765 @ 20?C (UK-sourced material)
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay from Supplier's CofA≥96.0% (US-sourced material)
Assay (GC)≥96.0% (UK-sourced material)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only