(-)-Shikimic acid, 98%
(-)-Shikimic acid, 98%
(-)-Shikimic acid, 98%
Thermo Scientific Chemicals

(-)-Shikimic acid, 98%

CAS: 138-59-0 | C7H9O5- | 173.144 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L04848.03
also known as L04848-03
Price (USD)
102.65
Special offer
Online exclusive
Ends: 30-Jun-2026
120.00
Save 17.35 (14%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
102.65
Special offer
Online exclusive
Ends: 30-Jun-2026
120.00
Save 17.35 (14%)
Each
Chemical Identifiers
CAS138-59-0
IUPAC Name3,4,5-trihydroxycyclohex-1-ene-1-carboxylic acid
Molecular FormulaC7H10O5
InChI KeyJXOHGGNKMLTUBP-UHFFFAOYNA-N
SMILESOC1CC(=CC(O)C1O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
Water Content (Karl Fischer Titration)≤1.0%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only