2-(Chloroethyl)benzene, 99%
2-(Chloroethyl)benzene, 99%
2-(Chloroethyl)benzene, 99%
Thermo Scientific Chemicals

2-(Chloroethyl)benzene, 99%

CAS: 622-24-2 | C8H9Cl | 140.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
500 g
Catalog number L04891.09
also known as L04891-09
Price (USD)
29.65
Online Exclusive
33.30
Save 3.65 (11%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
29.65
Online Exclusive
33.30
Save 3.65 (11%)
Each
Chemical Identifiers
CAS622-24-2
IUPAC Name(2-chloroethyl)benzene
Molecular FormulaC8H9Cl
InChI KeyMNNZINNZIQVULG-UHFFFAOYSA-N
SMILESClCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.5285-1.5315 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only