L-Adrenaline, 98+%
L-Adrenaline, 98+%
L-Adrenaline, 98+%
Thermo Scientific Chemicals

L-Adrenaline, 98+%

CAS: 51-43-4 | C9H13NO3 | 183.207 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L04911.06
also known as L04911-06
Price (USD)
72.65
Special offer
Online exclusive
Ends: 28-Dec-2026
84.80
Save 12.15
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
72.65
Special offer
Online exclusive
Ends: 28-Dec-2026
84.80
Save 12.15
Each
Chemical Identifiers
CAS51-43-4
IUPAC Name4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
Molecular FormulaC9H13NO3
InChI KeyUCTWMZQNUQWSLP-VIFPVBQESA-N
SMILESCNC[C@H](O)C1=CC=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0%
Optical Rotation-51.5 ± 2.0? (C=1 in 0.6N HCl)
Appearance (Color)White to cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only