3-Indoxyl acetate, 97%
3-Indoxyl acetate, 97%
3-Indoxyl acetate, 97%
Thermo Scientific Chemicals

3-Indoxyl acetate, 97%

CAS: 608-08-2 | C10H9NO2 | 175.187 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L04932.03
also known as L04932-03
Price (USD)
53.65
Online Exclusive
59.20
Save 5.55 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
53.65
Online Exclusive
59.20
Save 5.55 (9%)
Each
Chemical Identifiers
CAS608-08-2
IUPAC Name1H-indol-3-yl acetate
Molecular FormulaC10H9NO2
InChI KeyJBOPQACSHPPKEP-UHFFFAOYSA-N
SMILESCC(=O)OC1=CNC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream or pale yellow to pale brown or pale grey to pink
Assay (GC)≥96.0%
Useful for detection of esterases3-Indoxyl acetate acts as a colorimetric substrate used for differentiating esterases from various microbiological species. Further, it is used as a plant growth-stimulating hormone and reagent. In addition to this, it acts as a useful synthetic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful for detection of esterases3-Indoxyl acetate acts as a colorimetric substrate used for differentiating esterases from various microbiological species. Further, it is used as a plant growth-stimulating hormone and reagent. In addition to this, it acts as a useful synthetic intermediate.

Solubility
Soluble in ethanol, water, chloroform, dichloromethane and methanol.

Notes
Light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Substrate for enzyme assay: Anal. Chem., 37, 120 (1965).
  2. Darkoh, C.; Chappell, C.; Gonzales, C.; Okhuysen, P. A Rapid and Specific Method for the Detection of Indole in Complex Biological Samples. Appl. Environ. Microbiol. 2015, 81 (23), 8093-8097.
  3. Gandy, M. N.; Byrne, L. T.; Stubbs, K. A. A simple and robust preparation of N-acetylindoxyls: precursors for indigogenic substrates. Org. Biomol. Chem. 2015, 13 (3), 905-908.