4-Cyclohexylbenzoic acid, 98%
4-Cyclohexylbenzoic acid, 98%
4-Cyclohexylbenzoic acid, 98%
Thermo Scientific Chemicals

4-Cyclohexylbenzoic acid, 98%

CAS: 20029-52-1 | C13H16O2 | 204.269 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number L05086.04
also known as L05086-04
Price (USD)
45.65
Online Exclusive
50.60
Save 4.95 (10%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
45.65
Online Exclusive
50.60
Save 4.95 (10%)
Each
Chemical Identifiers
CAS20029-52-1
IUPAC Name4-cyclohexylbenzoic acid
Molecular FormulaC13H16O2
InChI KeyQCIWHVKGVVQHIY-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(C=C1)C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to red
FormCrystals or powder or crystalline powder or lumps or fused solid or granules
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only