1-Bromo-2-methylnaphthalene, tech. 90%
1-Bromo-2-methylnaphthalene, tech. 90%
1-Bromo-2-methylnaphthalene, tech. 90%
Thermo Scientific Chemicals

1-Bromo-2-methylnaphthalene, tech. 90%

CAS: 2586-62-1 | C11H9Br | 221.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L05093.18
also known as L05093-18
Price (USD)
100.65
Special offer
Online exclusive
Ends: 30-Jun-2026
118.00
Save 17.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
100.65
Special offer
Online exclusive
Ends: 30-Jun-2026
118.00
Save 17.35 (15%)
Each
Chemical Identifiers
CAS2586-62-1
IUPAC Name1-bromo-2-methylnaphthalene
Molecular FormulaC11H9Br
InChI KeyCMIMBQIBIZZZHQ-UHFFFAOYSA-N
SMILESCC1=CC=C2C=CC=CC2=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colourless or yellow to orange
Formliquid
Assay (GC)> 88.0%
Refractive Index1.6450 - 1.6530 @20?C
1-Bromo-2-methylnaphthalene is used as test compound in determination of halogenated hydrocarbons at trace levels by supercritical fluid chromatography-microwave-induced plasma mass spectrometry. Dynamic phosphorescence quenching of 1-bromo-2-methylnaphthalene without deoxygenation has been used for selective sensing of Cu(II) at ngm levels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-methylnaphthalene is used as test compound in determination of halogenated hydrocarbons at trace levels by supercritical fluid chromatography-microwave-induced plasma mass spectrometry. Dynamic phosphorescence quenching of 1-bromo-2-methylnaphthalene without deoxygenation has been used for selective sensing of Cu(II) at ngm levels.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
RUO – Research Use Only
Yu Wang; Jian-Jun Wu; Yu-Feng Wang; Li-Pin Qina and Wei-Jun Jin. Selective sensing of Cu(II) at ng ml-1 level based on phosphorescence quenching of 1-bromo-2-methylnaphthalene sandwiched in sodium deoxycholate dimer.Chem. Commun.2005, 1090-1091.