Glyoxime, 98+%, moistened with ca 20% water
Glyoxime, 98+%, moistened with ca 20% water
Glyoxime, 98+%, moistened with ca 20% water
Thermo Scientific Chemicals

Glyoxime, 98+%, moistened with ca 20% water

CAS: 557-30-2 | C2H4N2O2 | 88.066 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L05130.18
also known as L05130-18
Price (USD)
246.65
Online Exclusive
274.00
Save 27.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
246.65
Online Exclusive
274.00
Save 27.35 (10%)
Each
Chemical Identifiers
CAS557-30-2
IUPAC NameN-[(1E)-2-nitrosoethenyl]hydroxylamine
Molecular FormulaC2H4N2O2
InChI KeyRUFIRPVAEJIIIS-OWOJBTEDSA-N
SMILESONC=CN=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale pink or pale brown
Assay (HPLC)≥98.0%
FormCrystals or powder or crystalline powder or lumps or granules
Water Content (Karl Fischer Titration)17-23%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only