Isophthalamide, 98%
Isophthalamide, 98%
Isophthalamide, 98%
Thermo Scientific Chemicals

Isophthalamide, 98%

CAS: 1740-57-4 | C8H8N2O2 | 164.164 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L05137.14
also known as L05137-14
Price (USD)
112.00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
112.00
Each
Add to cart
Chemical Identifiers
CAS1740-57-4
IUPAC Namebenzene-1,3-dicarboxamide
Molecular FormulaC8H8N2O2
InChI KeyQZUPTXGVPYNUIT-UHFFFAOYSA-N
SMILESNC(=O)C1=CC(=CC=C1)C(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only