9-Aminofluorene hydrochloride, 98+%
9-Aminofluorene hydrochloride, 98+%
9-Aminofluorene hydrochloride, 98+%
9-Aminofluorene hydrochloride, 98+%
Thermo Scientific Chemicals

9-Aminofluorene hydrochloride, 98+%

CAS: 5978-75-6 | C13H12ClN | 217.70 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L05156.06
also known as L05156-06
Price (USD)
58.65
Special offer
Online exclusive
Ends: 30-Jun-2026
68.30
Save 9.65 (14%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
58.65
Special offer
Online exclusive
Ends: 30-Jun-2026
68.30
Save 9.65 (14%)
Each
Add to cart
Chemical Identifiers
CAS5978-75-6
IUPAC Namehydrogen 9H-fluoren-9-amine chloride
Molecular FormulaC13H12ClN
InChI KeySYKJOJSYQSVNOM-UHFFFAOYSA-N
SMILES[H+].[Cl-].NC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Chloride)≥98.0 to ≤102.0%
Appearance (Color)White to pale cream to cream to yellow to gray
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only