4-Chloro-2,5-dimethylbenzenesulfonyl chloride, 98%
4-Chloro-2,5-dimethylbenzenesulfonyl chloride, 98%
4-Chloro-2,5-dimethylbenzenesulfonyl chloride, 98%
Thermo Scientific Chemicals

4-Chloro-2,5-dimethylbenzenesulfonyl chloride, 98%

CAS: 88-49-3 | C8H8Cl2O2S | 239.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L05247.06
also known as L05247-06
Price (USD)
49.65
Special offer
Online exclusive
Ends: 30-Jun-2026
58.40
Save 8.75 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
49.65
Special offer
Online exclusive
Ends: 30-Jun-2026
58.40
Save 8.75 (15%)
Each
Chemical Identifiers
CAS88-49-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown or pale pink
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥97.5%
Melting Point (clear melt)45.0-52.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only