4-Chlorobenzylideneacetone, 98%
4-Chlorobenzylideneacetone, 98%
4-Chlorobenzylideneacetone, 98%
Thermo Scientific Chemicals

4-Chlorobenzylideneacetone, 98%

CAS: 3160-40-5 | C10H9ClO | 180.631 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L05321.06
also known as L05321-06
Price (USD)
45.65
Online exclusive
50.90
Save 5.25
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
45.65
Online exclusive
50.90
Save 5.25
Each
Specifications
Beilstein1100204
CAS3160-40-5
Chemical Name or Material4-Chlorobenzylideneacetone
EINECS Number221-610-5
Formula Weight180.64
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only