What regeneration temperature do you recommend for Molecular sieves, 3A, 1-2mm (0.04-0.08in) beads?
We recommend a regeneration temperature of 120-315 degrees Celsius for the Molecular sieves, 3A, 1-2mm (0.04-0.08in) beads.
Search
View additional product information for Molecular sieves, 3A, 1-2mm (0.04-0.08in) beads - FAQs (L05335.0I, L05335.30, L05335.0B)
1 product FAQs found