Ethyl 4-hydroxymandelate, 98%
Ethyl 4-hydroxymandelate, 98%
Ethyl 4-hydroxymandelate, 98%
Thermo Scientific Chemicals

Ethyl 4-hydroxymandelate, 98%

CAS: 68758-68-9 | C10H12O4 | 196.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L05413.03
also known as L05413-03
Price (USD)
28.65
Special offer
Online exclusive
Ends: 30-Jun-2026
33.10
Save 4.45 (13%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
28.65
Special offer
Online exclusive
Ends: 30-Jun-2026
33.10
Save 4.45 (13%)
Each
Chemical Identifiers
CAS68758-68-9
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White to cream
FormPowder or crystalline powder
Melting Point (clear melt)126-135?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only