Methyl palmitate, 97%
Methyl palmitate, 97%
Methyl palmitate, 97%
Thermo Scientific Chemicals

Methyl palmitate, 97%

CAS: 112-39-0 | C17H34O2 | 270.457 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number L05509.22
also known as L05509-22
Price (USD)
44.65
Online exclusive
49.50
Save 4.85
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
44.65
Online exclusive
49.50
Save 4.85
Each
Chemical Identifiers
CAS112-39-0
IUPAC Namemethyl hexadecanoate
Molecular FormulaC17H34O2
InChI KeyFLIACVVOZYBSBS-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)24.0-35.0?C
FormPowder or lumps or fused solid or waxy solid or liquid as melt
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Appearance (Color)Colorless to white to pale cream
Methyl palmitate is used in the preparation of detergents, emulsifiers, wetting agents, stabilizers, resins, lubricants, plasticizers and animal feeds. It exhibits an anti-inflammatory and anti-fibrotic agent and prevents bleomycin-induced lung inflammation and fibrosis in rats. In addition, it also prevents carbon tetrachloride-induced liver fibrosis linked to reduce transforming growth factor beta, which is a secreted protein that controls proliferation, cellular differentiation and other functions in most cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl palmitate is used in the preparation of detergents, emulsifiers, wetting agents, stabilizers, resins, lubricants, plasticizers and animal feeds. It exhibits an anti-inflammatory and anti-fibrotic agent and prevents bleomycin-induced lung inflammation and fibrosis in rats. In addition, it also prevents carbon tetrachloride-induced liver fibrosis linked to reduce transforming growth factor beta, which is a secreted protein that controls proliferation, cellular differentiation and other functions in most cells.

Solubility
Soluble in ethanol, chloroform, dimethyl sulfoxide and dimethyl formamide. Insoluble in water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Tang, B. K.; Lee, Y. J.; Row, K. H. Exploration of Using Deep Eutectic Solvents to Separate Methyl Palmitate from Simulated Biodiesel Mixtures. Adv. Mat. Res. 2015, 1101, 249-251.
  2. Wang, M.; Wu, W. W.; Wang, S. S.; Shi, X. Y.; Wu, F. A.; Wang, J. Preparation and Characterization of a Solid Acid Catalyst from Macro Fungi Residue for Methyl Palmitate Production. Bioresources 2015, 10 (3), 5691-5708.