2-Decyn-1-ol, 97%
2-Decyn-1-ol, 97%
2-Decyn-1-ol, 97%
Thermo Scientific Chemicals

2-Decyn-1-ol, 97%

CAS: 4117-14-0 | C10H18O | 154.253 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L05523.06
also known as L05523-06
Price (USD)
53.65
Special offer
Online exclusive
Ends: 30-Jun-2026
63.20
Save 9.55 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
53.65
Special offer
Online exclusive
Ends: 30-Jun-2026
63.20
Save 9.55 (15%)
Each
Specifications
Beilstein1852842
CAS4117-14-0
Chemical Name or Material2-Decyn-1-ol
Density0.855
EINECS Number000-000-0
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only