N,N-Dimethylbutyramide, 98%
N,N-Dimethylbutyramide, 98%
N,N-Dimethylbutyramide, 98%
N,N-Dimethylbutyramide, 98%
Thermo Scientific Chemicals

N,N-Dimethylbutyramide, 98%

CAS: 760-79-2 | C6H13NO | 115.176 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L05556.14
also known as L05556-14
Price (USD)
234.65
Online Exclusive
260.00
Save 25.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
234.65
Online Exclusive
260.00
Save 25.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS760-79-2
IUPAC NameN,N-dimethylbutanamide
Molecular FormulaC6H13NO
InChI KeyVIJUZNJJLALGNJ-UHFFFAOYSA-N
SMILESCCCC(=O)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4405-1.4445 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
Identification (FTIR)Conforms
N,N-Dimethylbutyramide, is used as an organic intermediate, pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylbutyramide, is used as an organic intermediate, pharmaceutical intermediate.

Solubility
Very Soluble in water (110 g/L) (25°C).

Notes
Stable under normal temperatures and pressures. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
RUO – Research Use Only

General References:

  1. HC Brown.; P Heim. Selective reductions. XVIII. Fast reaction of primary, secondary, and tertiary amides with diborane. Simple, convenient procedure for the conversion of amides to the corresponding amines. The Journal of Organic Chemistry. 1973, 38,(5), 912-916.
  2. HL Needles.; RE Whitfield. Alkylation of N, N-Dimethylamides via Carbanion Intermediates. The Journal of Organic Chemistry. 1966, 31,(3), 989-990.