Ethyl tert-butylacetate, 98%
Ethyl tert-butylacetate, 98%
Ethyl tert-butylacetate, 98%
Ethyl tert-butylacetate, 98%
Thermo Scientific Chemicals

Ethyl tert-butylacetate, 98%

CAS: 5340-78-3 | C8H16O2 | 144.214 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L05603.22
also known as L05603-22
Price (USD)
181.65
Online Exclusive
202.00
Save 20.35 (10%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
181.65
Online Exclusive
202.00
Save 20.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS5340-78-3
IUPAC Nameethyl 3,3-dimethylbutanoate
Molecular FormulaC8H16O2
InChI KeyJWMNHAMYTBAUPI-UHFFFAOYSA-N
SMILESCCOC(=O)CC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)Clear colorless
Refractive Index1.4015-1.4060 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only