Methyl 4-(methylsulfonylmethyl)benzoate, 98%
Methyl 4-(methylsulfonylmethyl)benzoate, 98%
Methyl 4-(methylsulfonylmethyl)benzoate, 98%
Thermo Scientific Chemicals

Methyl 4-(methylsulfonylmethyl)benzoate, 98%

CAS: 160446-22-0 | C10H12O4S | 228.262 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number L05657.04
also known as L05657-04
Price (USD)
67.65
Online Exclusive
75.20
Save 7.55 (10%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
67.65
Online Exclusive
75.20
Save 7.55 (10%)
Each
Chemical Identifiers
CAS160446-22-0
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥97.5%
FormCrystals or powder or crystalline powder
Appearance (Color)White
Melting Point (clear melt)161.5-167.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only