2-(4-Morpholinyl)aniline, 98%
2-(4-Morpholinyl)aniline, 98%
2-(4-Morpholinyl)aniline, 98%
Thermo Scientific Chemicals

2-(4-Morpholinyl)aniline, 98%

CAS: 5585-33-1 | C10H14N2O | 178.235 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L05660.14
also known as L05660-14
Price (USD)
225.65
Online Exclusive
250.00
Save 24.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
225.65
Online Exclusive
250.00
Save 24.35 (10%)
Each
Chemical Identifiers
CAS5585-33-1
IUPAC Name2-(morpholin-4-yl)aniline
Molecular FormulaC10H14N2O
InChI KeyQKWLVAYDAHQMLG-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1N1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or lumps
Appearance (Color)Pale cream to cream to brown or pale purple to purple to gray to black
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only