Thermo Scientific Chemicals

n-Nonadecane, 99%, Thermo Scientific Chemicals

Catalog number: L05701.14
25 g, Each
Thermo Scientific Chemicals

n-Nonadecane, 99%, Thermo Scientific Chemicals

Catalog number: L05701.14
25 g, Each
Quantity
Catalog number: L05701.14
also known as L05701-14
Price (USD)
Price: 33.70
Online price: 28.65
Your price:
Quantity
-

Chemical Identifiers

CAS
629-92-5
IUPAC Name
nonadecane
Molecular Formula
C19H40
InChI Key
LQERIDTXQFOHKA-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCCC
Appearance (Color)
White
Form
Crystalline, waxy or fused solid
Assay (GC)
≥98.5%
Melting Point (clear melt)
31-36?C

Description

n-Nonadecane is used as a fragrance agent in cosmetics and perfumes. It is used by egg parasitoid Trissolcus basalis to discriminate between female and male hosts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Nonadecane is used as a fragrance agent in cosmetics and perfumes. It is used by egg parasitoid Trissolcus basalis to discriminate between female and male hosts.

Solubility
Soluble in Chloroform, Hexane.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in well sealed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text