4-(Trifluoromethyl)benzhydrazide, 98%
4-(Trifluoromethyl)benzhydrazide, 98%
4-(Trifluoromethyl)benzhydrazide, 98%
Thermo Scientific Chemicals

4-(Trifluoromethyl)benzhydrazide, 98%

CAS: 339-59-3 | C8H7F3N2O | 204.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L05724.03
also known as L05724-03
Price (USD)
24.65
Online Exclusive
26.80
Save 2.15 (8%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
24.65
Online Exclusive
26.80
Save 2.15 (8%)
Each
Chemical Identifiers
CAS339-59-3
SpecificationsSpecification SheetSpecification Sheet
Melting point116-123°C (non-U.S. specification)
Purity≥97.5% (U.S. specification)
Appearance (Color)White to pale cream or pale yellow
FormPowder or crystalline powder or solid
Assay (HPLC)≥97.5% (non-U.S. specification)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only