2-Aminobiphenyl, 97%
2-Aminobiphenyl, 97%
2-Aminobiphenyl, 97%
Thermo Scientific Chemicals

2-Aminobiphenyl, 97%

CAS: 90-41-5 | C12H11N | 169.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L05728.06
also known as L05728-06
Price (USD)
93.65
Online Exclusive
104.00
Save 10.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
93.65
Online Exclusive
104.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS90-41-5
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%
Appearance (Color)White to brown
FormCrystals or Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only