1-Iodo-4-n-propylbenzene, 97%
1-Iodo-4-n-propylbenzene, 97%
1-Iodo-4-n-propylbenzene, 97%
Thermo Scientific Chemicals

1-Iodo-4-n-propylbenzene, 97%

CAS: 126261-84-5 | C9H11I | 246.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L05767.14
also known as L05767-14
Price (USD)
261.65
Special offer
Online exclusive
Ends: 30-Jun-2026
308.00
Save 46.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
261.65
Special offer
Online exclusive
Ends: 30-Jun-2026
308.00
Save 46.35 (15%)
Each
Chemical Identifiers
CAS126261-84-5
IUPAC Name1-iodo-4-propylbenzene
Molecular FormulaC9H11I
InChI KeyZRWCHMFGMKNWEC-UHFFFAOYSA-N
SMILESCCCC1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5760-1.5810 @ 20?C
FormLiquid
Appearance (Color)Clear colorless to yellow
Assay (GC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only