2,5-Dimethyl-1-phenylpyrrole-3-carboxaldehyde, 98+%
2,5-Dimethyl-1-phenylpyrrole-3-carboxaldehyde, 98+%
2,5-Dimethyl-1-phenylpyrrole-3-carboxaldehyde, 98+%
Thermo Scientific Chemicals

2,5-Dimethyl-1-phenylpyrrole-3-carboxaldehyde, 98+%

CAS: 83-18-1 | C13H13NO | 199.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L05836.06
also known as L05836-06
Price (USD)
93.65
Online Exclusive
104.00
Save 10.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
93.65
Online Exclusive
104.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS83-18-1
SpecificationsSpecification SheetSpecification Sheet
Free acid (titration)≤1.0%
Melting Point (clear melt)86.5-92.5?C
Appearance (Color)White to cream to pale brown
FormCrystals or powder or crystalline powder or lumpy
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only