Benzo[b]thiophene-3-acetic acid, 98+%
Benzo[b]thiophene-3-acetic acid, 98+%
Benzo[b]thiophene-3-acetic acid, 98+%
Thermo Scientific Chemicals

Benzo[b]thiophene-3-acetic acid, 98+%

CAS: 1131-09-5 | C10H8O2S | 192.232 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
2 g
10 g
Catalog number L05855.09
also known as L05855-09
Price (USD)
907.65
Online Exclusive
1,008.00
Save 100.35 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
907.65
Online Exclusive
1,008.00
Save 100.35 (10%)
Each
Chemical Identifiers
CAS1131-09-5
IUPAC Name2-(1-benzothiophen-3-yl)acetic acid
Molecular FormulaC10H8O2S
InChI KeyVFZQJKXVHYZXMM-UHFFFAOYSA-N
SMILESOC(=O)CC1=CSC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to dark cream to cream to yellow to pale brown
FormCrystals or powder or crystalline powder or fused solid or granules
Assay (HPLC)≥98.0%
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only