3-Indazolinone, 97%
3-Indazolinone, 97%
3-Indazolinone, 97%
Thermo Scientific Chemicals

3-Indazolinone, 97%

CAS: 7364-25-2 | C7H6N2O | 134.138 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L05860.06
also known as L05860-06
Price (USD)
70.65
Special offer
Online exclusive
Ends: 30-Jun-2026
83.30
Save 12.65 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
70.65
Special offer
Online exclusive
Ends: 30-Jun-2026
83.30
Save 12.65 (15%)
Each
Chemical Identifiers
CAS7364-25-2
IUPAC Name2,3-dihydro-1H-indazol-3-one
Molecular FormulaC7H6N2O
InChI KeySWEICGMKXPNXNU-UHFFFAOYSA-N
SMILESO=C1NNC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown or pale yellow
Assay (HPLC)≥96.0%
FormPowder
Melting Point (clear melt)239.0-248.0?C
3-Indazolinone is used as catalytic agent, petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Indazolinone is used as catalytic agent, petrochemical additive.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. S D Wyrick; P J Voorstad; G Cocolas; I H Hall. Hypolipidemic activity of phthalimide derivatives. 7. Structure-activity studies of indazolone analogues. Journal of Medicinal Chemistry. 1984, 27, (6), 768-772.
  2. V. M. F. Morais; M. S. Miranda; M. A. R. Matos & J. F Liebman. Experimental and computational thermochemistry of three nitrogen-containing heterocycles: 2-benzimidazolinone, 2-benzoxazolinone and 3-indazolinone.Molecular Physics: An International Journal at the Interface Between Chemistry and Physics. 2006, 104, 325-334.