Thermo Scientific Chemicals

3-Indazolinone, 97%, Thermo Scientific Chemicals

Catalog number: L05860.14
25 g, Each
Thermo Scientific Chemicals

3-Indazolinone, 97%, Thermo Scientific Chemicals

Catalog number: L05860.14
25 g, Each
Quantity
Catalog number: L05860.14
also known as L05860-14
Price (USD)
Price: 291.00
Online price: 249.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7364-25-2
IUPAC Name
2,3-dihydro-1H-indazol-3-one
Molecular Formula
C7H6N2O
InChI Key
SWEICGMKXPNXNU-UHFFFAOYSA-N
SMILES
O=C1NNC2=CC=CC=C12
Appearance (Color)
White to cream to pale brown or pale yellow
Assay (HPLC)
≥96.0%
Form
Powder
Melting Point (clear melt)
239.0-248.0?C

Description

3-Indazolinone is used as catalytic agent, petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Indazolinone is used as catalytic agent, petrochemical additive.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text