n-Decyltrichlorosilane, 97%
n-Decyltrichlorosilane, 97%
n-Decyltrichlorosilane, 97%
Thermo Scientific Chemicals

n-Decyltrichlorosilane, 97%

CAS: 13829-21-5 | C10H21Cl3Si | 275.713 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L05912.18
also known as L05912-18
Price (USD)
131.00
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
131.00
Each
Chemical Identifiers
CAS13829-21-5
IUPAC Nametrichloro(decyl)silane
Molecular FormulaC10H21Cl3Si
InChI KeyHLWCOIUDOLYBGD-UHFFFAOYSA-N
SMILESCCCCCCCCCC[Si](Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
Refractive Index1.4495-1.4545 @ 20?C
FormLiquid
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only