(+/-)-4-Phenyl-2-butanol, 98%
(+/-)-4-Phenyl-2-butanol, 98%
(+/-)-4-Phenyl-2-butanol, 98%
Thermo Scientific Chemicals

(+/-)-4-Phenyl-2-butanol, 98%

CAS: 2344-70-9 | C10H14O | 150.221 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L05970.06
also known as L05970-06
Price (USD)
47.40
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
47.40
Each
Chemical Identifiers
CAS2344-70-9
IUPAC Name4-phenylbutan-2-ol
Molecular FormulaC10H14O
InChI KeyGDWRKZLROIFUML-UHFFFAOYNA-N
SMILESCC(O)CCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Refractive Index1.5150-1.5190 @ 20°C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only