(+/-)-2-Hexanol, 99%
(+/-)-2-Hexanol, 99%
(+/-)-2-Hexanol, 99%
Thermo Scientific Chemicals

(+/-)-2-Hexanol, 99%

CAS: 626-93-7 | C6H14O | 102.177 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L05993.14
also known as L05993-14
Price (USD)
54.65
Special offer
Online exclusive
Ends: 30-Jun-2026
63.90
Save 9.25 (14%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
54.65
Special offer
Online exclusive
Ends: 30-Jun-2026
63.90
Save 9.25 (14%)
Each
Specifications
Beilstein1718996
CAS626-93-7
Chemical Name or Material(+/-)-2-Hexanol
DOT InformationTransport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HEXANOLS
Density0.81
View more
(±)-2-Hexanol is s an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff field. It is used as perfuming agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-2-Hexanol is s an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff field. It is used as perfuming agent.

Solubility
Soluble in water (13 g/L at 25°C),

Notes
Incompatible with acid chlorides and oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. Marıa Angeles Aramendıa; Victoriano Bora; César Jiménez; José Marıa Marinas; Francisco José Romero. Synthesis and Characterization of Magnesium Phosphates and Their Catalytic Properties in the Conversion of 2-Hexanol.Journal of Colloid and Interface Science.1999,217 288-298.
  2. María Angeles Aramendía; Victoriano Borau; César Jiménez; José María Marinas; Francisco José Romero. The Meerwein-Ponndorf-Verley-Oppenauer reaction between 2-hexanol and cyclohexanone on magnesium phosphates.Catalysis Letters.1999,58 53-58.