4'-Chloro-3'-methylacetophenone, tech. 75%, remainder 3'-chloro-4'-methylacetophenone
4'-Chloro-3'-methylacetophenone, tech. 75%, remainder 3'-chloro-4'-methylacetophenone
4'-Chloro-3'-methylacetophenone, tech. 75%, remainder 3'-chloro-4'-methylacetophenone
Thermo Scientific Chemicals

4'-Chloro-3'-methylacetophenone, tech. 75%, remainder 3'-chloro-4'-methylacetophenone

CAS: 37074-39-8 | C9H9ClO | 168.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L06011.06
also known as L06011-06
Price (USD)
56.65
Special offer
Online exclusive
Ends: 30-Jun-2026
66.40
Save 9.75 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
56.65
Special offer
Online exclusive
Ends: 30-Jun-2026
66.40
Save 9.75 (15%)
Each
Chemical Identifiers
CAS37074-39-8
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥70.0%
FormLiquid
Appearance (Color)Clear colorless to yellow
Refractive Index1.5505-1.5585 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only