2-(1-Pyrrolyl)benzoic acid, 99%
2-(1-Pyrrolyl)benzoic acid, 99%
2-(1-Pyrrolyl)benzoic acid, 99%
Thermo Scientific Chemicals

2-(1-Pyrrolyl)benzoic acid, 99%

CAS: 10333-68-3 | C11H9NO2 | 187.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L06069.06
also known as L06069-06
Price (USD)
168.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
168.00
Each
Chemical Identifiers
CAS10333-68-3
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Appearance (Color)Pale pink to cream to brown
Assay (HPLC)≥98.5%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only