4-Methoxyphenylhydrazine hydrochloride, 98%
4-Methoxyphenylhydrazine hydrochloride, 98%
4-Methoxyphenylhydrazine hydrochloride, 98%
Thermo Scientific Chemicals

4-Methoxyphenylhydrazine hydrochloride, 98%

CAS: 19501-58-7 | C7H11ClN2O | 174.628 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L06076.06
also known as L06076-06
Price (USD)
80.10
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
80.10
Each
Chemical Identifiers
CAS19501-58-7
IUPAC Name(4-methoxyphenyl)hydrazine hydrochloridyl
Molecular FormulaC7H10ClN2O
InChI KeyUNLQTHFXAIISEP-UHFFFAOYSA-N
SMILES[Cl].COC1=CC=C(NN)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Appearance (Color)White to pale pink to grey to purple
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only