Octaphenylcyclotetrasiloxane, 98+%
Octaphenylcyclotetrasiloxane, 98+%
Octaphenylcyclotetrasiloxane, 98+%
Thermo Scientific Chemicals

Octaphenylcyclotetrasiloxane, 98+%

CAS: 546-56-5 | C48H40O4Si4 | 793.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L06130.22
also known as L06130-22
Price (USD)
148.65
Online Exclusive
165.00
Save 16.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
148.65
Online Exclusive
165.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS546-56-5
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥98.0%
FormCrystalline powder
Melting Point (clear melt)196.0-203.0?C
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only